Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-trans-aconitic acid sodium salt hydrate 95.0 (TLC), ≥95% (TLC), MilliporeSigma™ Supelco™

Specifications

Percent Purity ≥95% (TLC)
Linear Formula C7H8O6 · xNa+ · yH2 O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-100-4934
View Documents
MilliporeSigma Supelco
1880210MG
10 mg N/A
11-100-4077
View Documents
MilliporeSigma Supelco
1880250MG
50 mg N/A
Description

Description

Specifications

Specifications

≥95% (TLC)
C7H8O6 · xNa+ · yH2 O
Safety and Handling

Safety and Handling

Recommended Storage : 2°C to 8°C

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit