Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Methylacetophenone 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
577-16-2
C9H10O
134.18
MFCD00008734
YXWWHNCQZBVZPV-UHFFFAOYSA-N
2'-methylacetophenone,1-o-tolyl ethanone,2-methylacetophenone,o-methylacetophenone,2-acetyltoluene,o-acetyltoluene,1-2-methylphenyl ethanone,ethanone, 1-2-methylphenyl,2'-methylacetylphenone,acetophenone, 2'-methyl
11340
1-(2-methylphenyl)ethanone
CC1=CC=CC=C1C(=O)C
577-16-2
C9H10O
134.18
MFCD00008734
YXWWHNCQZBVZPV-UHFFFAOYSA-N
2'-methylacetophenone,1-o-tolyl ethanone,2-methylacetophenone,o-methylacetophenone,2-acetyltoluene,o-acetyltoluene,1-2-methylphenyl ethanone,ethanone, 1-2-methylphenyl,2'-methylacetylphenone,acetophenone, 2'-methyl
11340
1-(2-methylphenyl)ethanone
CC1=CC=CC=C1C(=O)C
Boiling Point 214°C
Flash Point 75°C
Specific Gravity 1.026
Refractive Index 1.5290 to 1.533
Density 1.0260g/mL
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C6H4COCH3
Beilstein 07, 306
Formula Weight 134.18
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2'-Methylacetophenone, 98%
Show More Show Less

EINECSNumber : 209-408-5

TSCA : TSCA