Alfa Aesar™ 2-(Methylamino)butane hydrochloride, 94%

Manufacturer: Alfa Aesar™ H2754706

Quantity 5g

 View more versions of this product

Catalog No. AAH2754706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 75098-40-7
    Molecular Formula C5H13N·HCl
    Formula Weight 123.62
    MDL Number MFCD07777098
    Synonym 2-(Methylamino)butane hydrochloride
    Chemical Name or Material 2-(Methylamino)butane hydrochloride

    Specifications

    Formula Weight 123.62
    Molecular Formula C5H13N·HCl
    MDL Number MFCD07777098
    CAS 75098-40-7
    Synonym 2-(Methylamino)butane hydrochloride
    Health Hazard 1 Warning
    Melting Point 76° to 82°C
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 2-(Methylamino)butane hydrochloride
    Assay 94%

    Description

    Solubility
    Soluble in hydrochloric acid.

    Notes
    Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. 

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit