Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Methylamino)ethanol, 99%

Click to view available options
Quantity:
100mL
500mL
109-83-1
C3H9NO
75.111
MFCD00002839
OPKOKAMJFNKNAS-UHFFFAOYSA-N
2-methylamino ethanol,n-methylethanolamine,methylethanolamine,ethanol, 2-methylamino,methylethylolamine,n-methylaminoethanol,n-methylmonoethanolamine,n-monomethylethanolamine,2-hydroxyethyl methylamine,monomethylaminoethanol
8016
CHEBI:21763
2-(methylamino)ethanol
CNCCO
109-83-1
C3H9NO
75.111
MFCD00002839
OPKOKAMJFNKNAS-UHFFFAOYSA-N
2-methylamino ethanol,n-methylethanolamine,methylethanolamine,ethanol, 2-methylamino,methylethylolamine,n-methylaminoethanol,n-methylmonoethanolamine,n-monomethylethanolamine,2-hydroxyethyl methylamine,monomethylaminoethanol
8016
CHEBI:21763
2-(methylamino)ethanol
CNCCO
Applications
2-(Methylamino)ethanol is used as an intermediate in synthetic chemistry. It finds application in various fields such as textile lubricants, polishes, detergents and in personal care products. It is also involved in electrostatic automotive coatings and in acid gas scrubbing. Further, it serves as an intermediate in polymers and pharmaceuticals. In addition to this, it is used as a solvent in the natural gas-processing industry.

Solubility
Miscible with water, alcohol and ether.

Notes
Incompatible with oxidizing agents, acids, copper, copper alloys, zinc and galvanized iron.
Odor Ammonia-like
Quantity 500mL
Melting Point -5°C
Flash Point 74°C (165°F)
Boiling Point 159°C
Linear Formula CH3NH(CH2)2OH
UN Number UN2735
Beilstein 1071196
Merck Index 14,6015
Formula Weight 75.11
Percent Purity 99%
Chemical Name or Material 2-(Methylamino)ethanol
Show More Show Less

GHS H Statement
H314-H302-H312-H227
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

RTECSNumber : KL6650000

TSCA : TSCA