Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Methylamino)pyrimidine, 95%

Click to view available options
Quantity:
250mg
1g
931-61-3
C5H7N3
109.132
MFCD00234126
BQNXHDSGGRTFNX-UHFFFAOYSA-N
2-methylaminopyrimidine,2-pyrimidinamine, n-methyl,n-methyl-2-pyrimidinamine,methyl-pyrimidin-2-yl-amine,2-pyrimidinamine, n-methyl-9ci,methyl pyrimidin-2-yl amine,pyrimidine, 2-methylamino,methylaminopyrimidin,methyl-2 pyrimidinylamine,methylpyrimidin-2-ylamine
265776
N-methylpyrimidin-2-amine
CNC1=NC=CC=N1
931-61-3
C5H7N3
109.132
MFCD00234126
BQNXHDSGGRTFNX-UHFFFAOYSA-N
2-methylaminopyrimidine,2-pyrimidinamine, n-methyl,n-methyl-2-pyrimidinamine,methyl-pyrimidin-2-yl-amine,2-pyrimidinamine, n-methyl-9ci,methyl pyrimidin-2-yl amine,pyrimidine, 2-methylamino,methylaminopyrimidin,methyl-2 pyrimidinylamine,methylpyrimidin-2-ylamine
265776
N-methylpyrimidin-2-amine
CNC1=NC=CC=N1
Quantity 250mg
Melting Point 59°C to 64°C
Beilstein 109648
Formula Weight 109.13
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-(Methylamino)pyrimidine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning