Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylaminopyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
4597-87-9
C6H8N2
108.144
MFCD00006250
SVEUVITYHIHZQE-UHFFFAOYSA-N
2-methylamino pyridine,2-methylaminopyridine,2-pyridinamine, n-methyl,methylamino-2 pyridine,n-2-pyridylmethylamine,pyridine, 2-methylamino,n-methyl-2-pyridinamine,2-pyridinamine, n-methyl-9ci,n-methyl pyridimine
20727
N-methylpyridin-2-amine
CNC1=CC=CC=N1
4597-87-9
C6H8N2
108.144
MFCD00006250
SVEUVITYHIHZQE-UHFFFAOYSA-N
2-methylamino pyridine,2-methylaminopyridine,2-pyridinamine, n-methyl,methylamino-2 pyridine,n-2-pyridylmethylamine,pyridine, 2-methylamino,n-methyl-2-pyridinamine,2-pyridinamine, n-methyl-9ci,n-methyl pyridimine
20727
N-methylpyridin-2-amine
CNC1=CC=CC=N1
Color Colorless to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Density 1.05g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 108.14
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-Methylaminopyridine
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning