Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylanthracene 97%, Acros Organics

Click to view available options
Quantity:
250mg
613-12-7
C15H12
192.261
MFCD00003583
GYMFBYTZOGMSQJ-UHFFFAOYSA-N
anthracene, 2-methyl,anthracene, methyl,unii-6c868jov4a,ccris 2739,acmc-1bcf4,2-methylanthracene
11936
2-methylanthracene
CC1=CC2=CC3=CC=CC=C3C=C2C=C1
613-12-7
C15H12
192.261
MFCD00003583
GYMFBYTZOGMSQJ-UHFFFAOYSA-N
anthracene, 2-methyl,anthracene, methyl,unii-6c868jov4a,ccris 2739,acmc-1bcf4,2-methylanthracene
11936
2-methylanthracene
CC1=CC2=CC3=CC=CC=C3C=C2C=C1
Infrared Spectrum Authentic
Quantity 250mg
Melting Point 202°C to 206°C
Color Beige to Brown-Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 05, 674
Formula Weight 192.26
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-Methylanthracene, 97%
Show More Show Less