Alfa Aesar™ 2-Methylbenzeneboronic acid, 98%

Manufacturer: Alfa Aesar™ B2315403

Quantity 1g

 View more versions of this product

Catalog No. AAB2315403

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 16419-60-6
    Molecular Formula C7H9BO2
    Formula Weight 135.96
    MDL Number MFCD00093526
    Beilstein 2935796
    Synonym 2-Methylphenylboronic acid; o-Tolylboronic acid
    Chemical Name or Material 2-Methylbenzeneboronic acid

    Specifications

    Formula Weight 135.96
    Synonym 2-Methylphenylboronic acid; o-Tolylboronic acid
    Quantity 1g
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C7H9BO2
    MDL Number MFCD00093526
    CAS 16419-60-6
    Beilstein 2935796
    Health Hazard 1 Warning
    Melting Point ≈162° to 164°C
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number ED7777777
    Chemical Name or Material 2-Methylbenzeneboronic acid
    Assay 98%

    Description

    Applications
    2-Methylbenzeneboronic acid is a reagent used for Pd-catalyzed arylation using Suzuki-Miyaura cross-coupling in water, Ruthenium catalyzed direct arylation reactions, Ligand-free copper-catalyzed coupling reactions, Rhodium-catalyzed asymmetric 1,4-addition reactions. Reagent used in Preparation of chiral monophosphorus ligands in asymmetric Suzuki-Miyaura coupling reaction, Human farnesyl pyrophosphate synthase inhibitors as antitumor agents for multiple myeloma cells

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit