Alfa Aesar™ 2-Methylbenzhydrol, 98+%

Manufacturer: Alfa Aesar™ B2302136

Quantity 500g

 View more versions of this product

Catalog No. AAB2302136

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5472-13-9
    Molecular Formula C14H14O
    Formula Weight 198.27
    MDL Number MFCD00004490
    Beilstein 1952703
    Synonym 2-Methyldiphenylmethanol; Phenyl o-tolyl carbinol
    Chemical Name or Material 2-Methylbenzhydrol

    Specifications

    Formula Weight 198.27
    TSCA TSCA
    Molecular Formula C14H14O
    MDL Number MFCD00004490
    CAS 5472-13-9
    Beilstein 1952703
    Synonym 2-Methyldiphenylmethanol; Phenyl o-tolyl carbinol
    Boiling Point 323°C
    Melting Point 93° to 95°C
    Quantity 500g
    Chemical Name or Material 2-Methylbenzhydrol
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit