Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbenzofuran, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
4265-25-2
C9H8O
132.16
GBGPVUAOTCNZPT-UHFFFAOYSA-N
2-methylbenzofuran,benzofuran, 2-methyl,2-methylcumarone,2-methylbenzo b furan,2-methyl benzofuran,unii-8x3183bz3x,methylbenzofuran,methyl benzoxole,2-methyl-benzofuran,#
20263
2-methyl-1-benzofuran
CC1=CC2=CC=CC=C2O1
4265-25-2
C9H8O
132.16
GBGPVUAOTCNZPT-UHFFFAOYSA-N
2-methylbenzofuran,benzofuran, 2-methyl,2-methylcumarone,2-methylbenzo b furan,2-methyl benzofuran,unii-8x3183bz3x,methylbenzofuran,methyl benzoxole,2-methyl-benzofuran,#
20263
2-methyl-1-benzofuran
CC1=CC2=CC=CC=C2O1
Flash Point 67°C
Color Colorless to Yellow
Packaging Glass bottle
Quantity 5g
Boiling Point 197.0°C to 198.0°C
Density 1.0570g/mL
Refractive Index 1.5618
Specific Gravity 1.057
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Beilstein 17,6
Formula Weight 132.16
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 2-Methylbenzofuran
Show More Show Less

GHS H Statement:

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

EINECSNumber : 224-249-1