Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbenzothiazole 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
120-75-2
C8H7NS
149.21
DXYYSGDWQCSKKO-UHFFFAOYSA-N
2-methylbenzothiazole,benzothiazole, 2-methyl,2-methybenzothiazole,2-methyl-benzothiazole,benzothiazole, methyl,usaf ek-1853,unii-7lx1xe6h8w,2-methylbenzo d thiazole,2-methyl benzothiazole,7lx1xe6h8w
8446
2-methyl-1,3-benzothiazole
CC1=NC2=CC=CC=C2S1
120-75-2
C8H7NS
149.21
DXYYSGDWQCSKKO-UHFFFAOYSA-N
2-methylbenzothiazole,benzothiazole, 2-methyl,2-methybenzothiazole,2-methyl-benzothiazole,benzothiazole, methyl,usaf ek-1853,unii-7lx1xe6h8w,2-methylbenzo d thiazole,2-methyl benzothiazole,7lx1xe6h8w
8446
2-methyl-1,3-benzothiazole
CC1=NC2=CC=CC=C2S1
Refractive Index 1.6150 to 1.6180
Infrared Spectrum Authentic
Specific Gravity 1.2
Melting Point 12.0°C to 14.0°C
Boiling Point 238.0°C
Packaging Glass bottle
Color Amber to Colorless
Quantity 100g
Density 1.2000g/mL
Flash Point 102°C
CAS Min % 98.5
CAS Max % 99.0
Assay Percent Range 99%
Beilstein 27,46
Solubility Information Solubility in water: insoluble. Other solubilities: ssluble in ether and chloroform
Formula Weight 149.21
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2-Methylbenzothiazole, 99%
Show More Show Less

GHS P Statement
Harmful if swallowed.

GHS P Statement
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 204-423-3

RTECSNumber : DL5600000

TSCA : TSCA