Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbenzyl Cyanide 98%, Acros Organics

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
22364-68-7
C9H9N
131.178
MFCD00001904
WMGVPDQNPUQRND-UHFFFAOYSA-N
2-methylbenzyl cyanide,2-o-tolyl acetonitrile,o-tolylacetonitrile,2-methylphenylacetonitrile,o-methylphenylacetonitrile,o-xylyl cyanide,o-methylbenzyl cyanide,acetonitrile, o-tolyl,benzeneacetonitrile, 2-methyl,2-2-methylphenyl acetonitrile
31155
CHEBI:27982
2-(2-methylphenyl)acetonitrile
CC1=CC=CC=C1CC#N
22364-68-7
C9H9N
131.178
MFCD00001904
WMGVPDQNPUQRND-UHFFFAOYSA-N
2-methylbenzyl cyanide,2-o-tolyl acetonitrile,o-tolylacetonitrile,2-methylphenylacetonitrile,o-methylphenylacetonitrile,o-xylyl cyanide,o-methylbenzyl cyanide,acetonitrile, o-tolyl,benzeneacetonitrile, 2-methyl,2-2-methylphenyl acetonitrile
31155
CHEBI:27982
2-(2-methylphenyl)acetonitrile
CC1=CC=CC=C1CC#N
Color Colorless to Orange-Red
Infrared Spectrum Authentic
Density 1.05g/mL
Packaging Glass bottle
Refractive Index 1.5260 to 1.5280 (20°C, 589nm)
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C6H4CH2CN
Beilstein 09, 527
Formula Weight 131.18
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-Methylbenzyl cyanide, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning