Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methylbenzyl mercaptan, 97%

Click to view available options
Quantity:
5g
25g
100g
7341-24-4
C8H10S
138.228
MFCD00022074
PJUDFYDAJBQPEA-UHFFFAOYSA-N
2-methylphenyl methanethiol,2-methylbenzyl mercaptan,2-methylbenzylmercaptan,2-methylbenzenemethanethiol,o-methylbenzyl mercaptan,o-tolylmethanethiol,o-tolyl-methanethiol,2-methylphenyl methane-1-thiol
288028
(2-methylphenyl)methanethiol
CC1=CC=CC=C1CS
7341-24-4
C8H10S
138.228
MFCD00022074
PJUDFYDAJBQPEA-UHFFFAOYSA-N
2-methylphenyl methanethiol,2-methylbenzyl mercaptan,2-methylbenzylmercaptan,2-methylbenzenemethanethiol,o-methylbenzyl mercaptan,o-tolylmethanethiol,o-tolyl-methanethiol,2-methylphenyl methane-1-thiol
288028
(2-methylphenyl)methanethiol
CC1=CC=CC=C1CS
Sensitivity Air Sensitive
Refractive Index 1.572
Odor Stench
Boiling Point 56°C to 58°C (5mmHg)
Quantity 100g
UN Number UN2810
Beilstein 2040956
Formula Weight 138.23
Percent Purity 97%
Chemical Name or Material 2-Methylbenzyl mercaptan

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen