Alfa Aesar™ (2-Methylbenzyl)triphenylphosphonium chloride, 98+%

Manufacturer: Alfa Aesar™ A1071918

Quantity 50g

 View more versions of this product

Catalog No. AAA1071918

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 63368-36-5
    Molecular Formula C26H24ClP
    Formula Weight 402.91
    MDL Number MFCD00031577
    Beilstein 4172676
    Chemical Name or Material (2-Methylbenzyl)triphenylphosphonium chloride

    Specifications

    Formula Weight 402.91
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00031577
    Beilstein 4172676
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    Molecular Formula C26H24ClP
    CAS 63368-36-5
    Health Hazard 1 Warning
    Melting Point 270° to 275°C
    Quantity 50g
    Chemical Name or Material (2-Methylbenzyl)triphenylphosphonium chloride
    Assay ≥98%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit