Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbenzylmagnesium chloride, 0.25M solution in THF, AcroSeal™, Acros Organics

Click to view available options
Quantity:
50mL
Packaging:
AcroSeal™ Glass Bottle
29875-05-6,109-99-9
C8H9ClMg
164.92
YGJUEZWNAWCXGJ-UHFFFAOYSA-M
chloro 2-methylphenyl methyl magnesium,magnesium,chloro 2-methylphenyl methyl,magnesium, chloro 2-methylphenyl methyl,2-methylbenzylmagnesium chloride solution,o-methylbenzylmagnesium chloride,2-methylbenzylmagnesium chloride,2-methylbenzylmagnesium chloride, 0.25m in 2-methf,2-methylbenzylmagnesium chloride 0.25 m in tetrahydrofuran
122498
magnesium;1-methanidyl-2-methylbenzene;chloride
CC1=CC=CC=C1[CH2-].[Mg+2].[Cl-]
29875-05-6,109-99-9
C8H9ClMg
164.92
YGJUEZWNAWCXGJ-UHFFFAOYSA-M
chloro 2-methylphenyl methyl magnesium,magnesium,chloro 2-methylphenyl methyl,magnesium, chloro 2-methylphenyl methyl,2-methylbenzylmagnesium chloride solution,o-methylbenzylmagnesium chloride,2-methylbenzylmagnesium chloride,2-methylbenzylmagnesium chloride, 0.25m in 2-methf,2-methylbenzylmagnesium chloride 0.25 m in tetrahydrofuran
122498
magnesium;1-methanidyl-2-methylbenzene;chloride
CC1=CC=CC=C1[CH2-].[Mg+2].[Cl-]
Density 0.9196g/mL
Quantity 50mL
Packaging AcroSeal™ Glass Bottle
Color Gray to Yellow
Flash Point −17°C
Specific Gravity 0.9196
CAS Min % 3.95,94.98
CAS Max % 5.02,96.05
Formula Weight 164.92
Physical Form Solution
Chemical Name or Material 2-Methylbenzylmagnesium chloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Reacts violently with water.
May form explosive peroxides.
Suspected of causing cancer.<br/

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger