Learn More
2-Methylbutane, 99+%, Thermo Scientific Chemicals
Description
2-Methylbutane is used as a chemical intermediate. It acts as a blowing agent for polystyrene and gasoline additive. It is used as a solvent of polyethylene and involved in the preparation of polystyrene foam and polyurethane foam. Further, it is used in a closed loop in geothermal power production to operate turbines. In addition to this, it is used to freeze biological samples like tissues in place of dry ice for cryosectioning in histology.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 0.62 |
Boiling Point | 30°C |
Flash Point | -57°C (-70°F) |
Assay Percent Range | 99+% |
Refractive Index | 1.354 |
Quantity | 500 mL |
Solubility Information | Miscible with water,hydrocarbons,oils,alcohol and ether. |
Formula Weight | 72.14 |
Physical Form | Liquid |
Chemical Name or Material | 2-Methylbutane |
Safety and Handling
Hazard Category | H224-H304-H336 |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c |
DOTInformation | Hazard Class: 3; Packaging Group: I |
EINECSNumber | 201-142-8 |
RTECSNumber | EK4430000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.