Alfa Aesar™ 2-Methylbutyraldehyde, 98%

Manufacturer: Alfa Aesar™ 31855-36

Quantity 500g

 View more versions of this product

Catalog No. AA3185536

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 96-17-3
    Formula Weight 86.13
    MDL Number MFCD00006984
    UN Number UN1989
    Synonym 2-Methylbutanal
    Chemical Name or Material 2-Methylbutyraldehyde

    Specifications

    Formula Weight 86.13
    Density 0.8030g/mL
    Physical Form Liquid
    Flash Point 4° (39°F)
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately al
    UN Number UN1989
    Synonym 2-Methylbutanal
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALDEHYDES, N.O.S.
    Refractive Index 1.3919
    CAS 96-17-3
    Boiling Point 90° to 92°C
    Health Hazard 1 Danger
    Quantity 500g
    TSCA TSCA
    MDL Number MFCD00006984
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 2-Methylbutyraldehyde
    Assay 98%

    GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately al

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit