2-Methylbutyric acid, 98%

Manufacturer: Alfa Aesar A11546AE

Quantity 100mL

 View more versions of this product

Catalog No. AAA11546AE

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 116-53-0
    Molecular Formula CH3CH2CH(CH3)CO2H
    Formula Weight 102.14
    MDL Number MFCD00002669
    UN Number UN3265
    Beilstein 1720486
    Chemical Name or Material 2-Methylbutyric Acid

    Specifications

    Formula Weight 102.14
    Quantity 100mL
    TSCA TSCA
    MDL Number MFCD00002669
    UN Number UN3265
    Beilstein 1720486
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H314-H318-H227
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON S
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    RTECS Number EK7897000
    Physical Form Liquid
    Density 0.937
    Molecular Formula CH3CH2CH(CH3)CO2H
    CAS 116-53-0
    Assay Percent Range 98%
    Chemical Name or Material 2-Methylbutyric Acid
    Assay 98%

    Description

    Applications
    2-Methylbutyric acid, can be used as food additive for butter, cream, cheese flavor deployment. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used in the synthesis of 2-methylbutyric anhydride.

    Solubility
    It is soluble in water, but highly soluble in most common organic solvents such as Chloroform, Methanol.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Bases, Oxidizing agents, Reducing agents.

    GHS H Statement
    H301-H311-H314-H318-H227
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON S

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit