Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylcyclohexanone, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
50g
100g
250g
500g
1kg
583-60-8
C7H12O
112.17
LFSAPCRASZRSKS-UHFFFAOYSA-N
2-methylcyclohexanone,o-methylcyclohexanone,cyclohexanone, 2-methyl,2-metilcicloesanone,2-methyl-1-cyclohexanone,methylanon,alpha-methylcyclohexanone,sexton b,2-methyl-cyclohexanon,2-methyl cyclohexanone
11419
2-methylcyclohexan-1-one
CC1CCCCC1=O
583-60-8
C7H12O
112.17
LFSAPCRASZRSKS-UHFFFAOYSA-N
2-methylcyclohexanone,o-methylcyclohexanone,cyclohexanone, 2-methyl,2-metilcicloesanone,2-methyl-1-cyclohexanone,methylanon,alpha-methylcyclohexanone,sexton b,2-methyl-cyclohexanon,2-methyl cyclohexanone
11419
2-methylcyclohexan-1-one
CC1CCCCC1=O
Specific Gravity 0.924
Flash Point 48°C
Color Colorless to Yellow
Packaging Glass bottle
Boiling Point 162.0°C to 163.0°C
Melting Point -14.0°C
Density 0.9240g/mL
Refractive Index 1.4470 to 1.4490
Infrared Spectrum Authentic
Quantity 50g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3C6H9(=O)
Beilstein 07, 14
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 112.17
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Methylcyclohexanone, 98%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-513-6

RTECSNumber : GW1750000

TSCA : TSCA