Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylcyclohexylamine, 98%, mixture of cis and trans, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
7003-32-9
C7H15N
113.204
MFCD00001492
FEUISMYEFPANSS-UHFFFAOYSA-N
2-methylcyclohexylamine,cyclohexanamine, 2-methyl,o-methylcyclohexylamine,cyclohexylamine, 2-methyl,2-methylcyclohexanamine,2-methylcyclohexylamine,c&t,hexahydro-o-toluidine,2methylcyclohexylamine,2-methyl-cyclohexylamine,2-methylcyclohexyl amine
23432
2-methylcyclohexan-1-amine
CC1CCCCC1N
7003-32-9
C7H15N
113.204
MFCD00001492
FEUISMYEFPANSS-UHFFFAOYSA-N
2-methylcyclohexylamine,cyclohexanamine, 2-methyl,o-methylcyclohexylamine,cyclohexylamine, 2-methyl,2-methylcyclohexanamine,2-methylcyclohexylamine,c&t,hexahydro-o-toluidine,2methylcyclohexylamine,2-methyl-cyclohexylamine,2-methylcyclohexyl amine
23432
2-methylcyclohexan-1-amine
CC1CCCCC1N
Quantity 10g
Density 0.8500g/mL
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Refractive Index 1.4555 to 1.4575 (20°C, 589nm)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C6H10NH2
Beilstein 12, 9
Formula Weight 113.2
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-Methylcyclohexylamine, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 230-277-5

TSCA : TSCA