Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylcyclopropanemethanol, 98%, mixture of cis and trans, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass Bottle
6077-72-1
C5H10O
86.134
MFCD00009683
SHEINYPABNPRPM-UHFFFAOYSA-N
2-methylcyclopropyl methanol,2-methylcyclopropanemethanol,cyclopropanemethanol, 2-methyl,2-methylcyclopropanemethanol, mixture of cis and trans,acmc-20a6ci,2-methyl-cyclopropane-methanol,cyclopropanemethanol,2-methyl,2-methylcyclopropyl methan-1-ol,2-methylcyclopropanemethanol, cis + trans
138655
(2-methylcyclopropyl)methanol
CC1CC1CO
6077-72-1
C5H10O
86.134
MFCD00009683
SHEINYPABNPRPM-UHFFFAOYSA-N
2-methylcyclopropyl methanol,2-methylcyclopropanemethanol,cyclopropanemethanol, 2-methyl,2-methylcyclopropanemethanol, mixture of cis and trans,acmc-20a6ci,2-methyl-cyclopropane-methanol,cyclopropanemethanol,2-methyl,2-methylcyclopropyl methan-1-ol,2-methylcyclopropanemethanol, cis + trans
138655
(2-methylcyclopropyl)methanol
CC1CC1CO
Specific Gravity 0.87
Flash Point 39°C
Boiling Point 133.0°C
Color Colorless
Quantity 10g
Packaging Glass Bottle
Refractive Index 1.4280 to 1.4330
Infrared Spectrum Authentic
Density 0.8700g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C3H4CH2OH
Formula Weight 86.13
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Methylcyclopropanemethanol, 98%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA