Alfa Aesar™ 2-Methylcyclopropanemethanol, cis + trans, 98%

Manufacturer: Alfa Aesar™ H2733709

Quantity 10g

 View more versions of this product

Catalog No. AAH2733709

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6077-72-1
    Molecular Formula C5H10O
    Formula Weight 86.13
    MDL Number MFCD00009683
    UN Number UN1987
    Chemical Name or Material 2-Methylcyclopropanemethanol, Cis + Trans

    Specifications

    Formula Weight 86.13
    Molecular Formula C5H10O
    UN Number UN1987
    Flash Point 39°C (102°F)
    Odor Alcohol-like
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00009683
    CAS 6077-72-1
    Health Hazard 1 Warning
    Boiling Point 133°C
    Quantity 10g
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
    Chemical Name or Material 2-Methylcyclopropanemethanol, Cis + Trans
    Assay 98%

    Description

    Applications
    It is employed in the radical clock investigation of microsomal cytochrome P-450 hydroxylation of hydrocarbons.

    Solubility
    Slightly soluble in water.

    Notes
    Store away from halogens and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away and protect from electrostatic charges.

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit