Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methylformanilide, 98%

Click to view available options
Quantity:
5g
25g
100g
94-69-9
C8H9NO
135.166
MFCD00014122
ZXTLGJAARBNQGK-UHFFFAOYSA-N
2'-methylformanilide,2-methylformanilide,o-tolylformamide,n-2-methylphenyl formamide,formamide, n-2-methylphenyl,o-methylformanilide,o-formotoluidide,o-methyl-n-formylaniline,2-methylphenylformamide,n-formyl-2-methylaniline
7202
N-(2-methylphenyl)formamide
CC1=CC=CC=C1NC=O
94-69-9
C8H9NO
135.166
MFCD00014122
ZXTLGJAARBNQGK-UHFFFAOYSA-N
2'-methylformanilide,2-methylformanilide,o-tolylformamide,n-2-methylphenyl formamide,formamide, n-2-methylphenyl,o-methylformanilide,o-formotoluidide,o-methyl-n-formylaniline,2-methylphenylformamide,n-formyl-2-methylaniline
7202
N-(2-methylphenyl)formamide
CC1=CC=CC=C1NC=O
Boiling Point 287°C to 289°C
Quantity 25g
Melting Point 58°C to 60°C
Beilstein 2205748
Formula Weight 135.17
Percent Purity 98%
Chemical Name or Material 2-Methylformanilide

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA