Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylformanilide, 98%, Acros Organics

Click to view available options
Quantity:
5g
94-69-9
C8H9NO
135.166
MFCD00014122
ZXTLGJAARBNQGK-UHFFFAOYSA-N
2'-methylformanilide,2-methylformanilide,o-tolylformamide,n-2-methylphenyl formamide,formamide, n-2-methylphenyl,o-methylformanilide,o-formotoluidide,o-methyl-n-formylaniline,2-methylphenylformamide,n-formyl-2-methylaniline
7202
N-(2-methylphenyl)formamide
CC1=CC=CC=C1NC=O
94-69-9
C8H9NO
135.166
MFCD00014122
ZXTLGJAARBNQGK-UHFFFAOYSA-N
2'-methylformanilide,2-methylformanilide,o-tolylformamide,n-2-methylphenyl formamide,formamide, n-2-methylphenyl,o-methylformanilide,o-formotoluidide,o-methyl-n-formylaniline,2-methylphenylformamide,n-formyl-2-methylaniline
7202
N-(2-methylphenyl)formamide
CC1=CC=CC=C1NC=O
Color Beige
Infrared Spectrum Authentic
Quantity 5g
Melting Point 57°C to 61°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula OCHNHC6H4(CH3)
Formula Weight 135.17
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Methylformanilide, 98%
Show More Show Less