Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylfuran-3-thiolacetate, 80%, Acros Organics

Click to view available options
Quantity:
1g
5g
55764-25-5
C7H8O2S
156.199
MFCD01632595
PQFIBPDAGFGLBY-UHFFFAOYSA-N
2-methylfuran-3-thiol acetate,3-acetylthio-2-methylfuran,ethanethioic acid, s-2-methyl-3-furanyl ester,s-2-methyl-3-furyl thioacetate,s-2-methyl-3-furyl ethanethioate,2-methyl-3-furanthiol, acetate,unii-7277jee7af,s-2-methylfuran-3-yl ethanethioate,thioacetic acid s-2-methyl-3-furyl ester,1-2-methylfuran-3-yl sulfanyl ethanone
108765
S-(2-methylfuran-3-yl) ethanethioate
CC1=C(C=CO1)SC(=O)C
55764-25-5
C7H8O2S
156.199
MFCD01632595
PQFIBPDAGFGLBY-UHFFFAOYSA-N
2-methylfuran-3-thiol acetate,3-acetylthio-2-methylfuran,ethanethioic acid, s-2-methyl-3-furanyl ester,s-2-methyl-3-furyl thioacetate,s-2-methyl-3-furyl ethanethioate,2-methyl-3-furanthiol, acetate,unii-7277jee7af,s-2-methylfuran-3-yl ethanethioate,thioacetic acid s-2-methyl-3-furyl ester,1-2-methylfuran-3-yl sulfanyl ethanone
108765
S-(2-methylfuran-3-yl) ethanethioate
CC1=C(C=CO1)SC(=O)C
Quantity 1g
Refractive Index 1.5140 to 1.5160 (20°C, 589nm)
Color Colorless to Brown
Infrared Spectrum Authentic
CAS Min % 75.0
CAS Max % 100.0
Assay Percent Range 80%
Formula Weight 156.2
Physical Form Liquid
Percent Purity ≥75%
Chemical Name or Material 2-Methylfuran-3-thiolacetate, 80%
Show More Show Less

STENCH

EINECSNumber : 259-801-0

TSCA : TSCA