Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylheptane 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
592-27-8
C8H18
114.23
MFCD00009518
JVSWJIKNEAIKJW-UHFFFAOYSA-N
heptane, 2-methyl,methylheptane,2-methyl-heptane,unii-yu6su8ccvb,yu6su8ccvb,hexane, dimethyl,dimethylhexane,acmc-1avkm,2-methylheptane,ch3 2ch ch2 4ch3
11594
2-methylheptane
CCCCCC(C)C
592-27-8
C8H18
114.23
MFCD00009518
JVSWJIKNEAIKJW-UHFFFAOYSA-N
heptane, 2-methyl,methylheptane,2-methyl-heptane,unii-yu6su8ccvb,yu6su8ccvb,hexane, dimethyl,dimethylhexane,acmc-1avkm,2-methylheptane,ch3 2ch ch2 4ch3
11594
2-methylheptane
CCCCCC(C)C
Melting Point -108.9°C
Boiling Point 116°C (761.0mmHg)
Flash Point 4°C
Specific Gravity 0.69
Density 0.6900g/mL
Refractive Index 1.3940 to 1.396
Packaging Glass bottle
Quantity 25g
Color Colorless
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3(CH2)4CH(CH3)2
Beilstein 01, 161
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether,miscible with ethanol,acetone
Formula Weight 114.23
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 2-Methylheptane, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-747-9

TSCA : TSCA