Alfa Aesar™ 2-Methylhexanoic acid, 98%

Manufacturer: Alfa Aesar™ A1129022

Quantity 100g

 View more versions of this product

Catalog No. AAA1129022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4536-23-6
    Molecular Formula C7H14O2
    Formula Weight 130.19
    MDL Number MFCD00002674
    UN Number UN3265
    Beilstein 1721227
    Synonym 2-Methylcaproic acid
    Chemical Name or Material 2-Methylhexanoic acid

    Specifications

    Formula Weight 130.19
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C7H14O2
    CAS 4536-23-6
    Synonym 2-Methylcaproic acid
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    TSCA TSCA
    MDL Number MFCD00002674
    UN Number UN3265
    Beilstein 1721227
    Health Hazard 1 Danger
    RTECS Number MO8400600
    Flash Point 105°C (221°F)
    Odor Rancid
    Boiling Point 209° to 210°C
    Quantity 100g
    Chemical Name or Material 2-Methylhexanoic acid
    Assay 98%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit