Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylhydroquinone, 99%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
95-71-6
C7H8O2
124.14
MFCD00002345
CNHDIAIOKMXOLK-UHFFFAOYSA-N
methylhydroquinone,2-methylhydroquinone,2,5-dihydroxytoluene,p-toluhydroquinone,toluhydroquinone,toluquinol,p-toluquinol,tolylhydroquinone,p-toluhydroquinol,2,5-toluenediol
7253
2-methylbenzene-1,4-diol
CC1=C(C=CC(=C1)O)O
95-71-6
C7H8O2
124.14
MFCD00002345
CNHDIAIOKMXOLK-UHFFFAOYSA-N
methylhydroquinone,2-methylhydroquinone,2,5-dihydroxytoluene,p-toluhydroquinone,toluhydroquinone,toluquinol,p-toluquinol,tolylhydroquinone,p-toluhydroquinol,2,5-toluenediol
7253
2-methylbenzene-1,4-diol
CC1=C(C=CC(=C1)O)O
Flash Point 172°C
Color Beige to Gray-White
Melting Point 126.0°C
Infrared Spectrum Authentic
Quantity 1kg
Packaging Plastic bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 06, 874
Solubility Information Solubility in water: 77g/L (25°C). Other solubilities: soluble in alcohol,benzene and chloroform
Formula Weight 124.14
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 2-Methylhydroquinone
Show More Show Less

GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.
Suspected of causing genetic defects.
Suspected of causing cancer.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.