Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylnaphthalene-bis(hexachlorocyclopentadiene) adduct, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Plastic Bottle
4605-91-8
C21H10Cl12
687.711
MFCD00167974
IBGAOSYQYASNKV-REWQHHMFSA-N
2-methylnaphthalene-bis hexachlorocyclopentadiene adduct
131879471
CC1=CC2=C(C=C1)C3C(C4C2C5(C(=C(C4(C5(Cl)Cl)Cl)Cl)Cl)Cl)C6(C(=C(C3(C6(Cl)Cl)Cl)Cl)Cl)Cl
4605-91-8
C21H10Cl12
687.711
MFCD00167974
IBGAOSYQYASNKV-REWQHHMFSA-N
2-methylnaphthalene-bis hexachlorocyclopentadiene adduct
131879471
CC1=CC2=C(C=C1)C3C(C4C2C5(C(=C(C4(C5(Cl)Cl)Cl)Cl)Cl)Cl)C6(C(=C(C3(C6(Cl)Cl)Cl)Cl)Cl)Cl
Quantity 25g
Color White to Beige
Melting Point 160°C to 165°C
Packaging Plastic Bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 687.75
Physical Form Amorphous Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-Methylnaphthalene-bis(hexachlorocyclopentadiene) adduct, 97%
Show More Show Less