Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylnaphtho[1,2-d]thiazole 98.0+%, TCI America™

Click to view available options
Quantity:
5g
10g
2682-45-3
C12H9NS
199.271
MFCD00004951
OUXMJRMYZCEVKO-UHFFFAOYSA-N
2-methylnaphtho 1,2-d thiazole,2-methyl-beta-naphthothiazole,2-methylnaphtho 1,2-d 1,3 thiazole,naphtho 1,2-d thiazole, 2-methyl,2-methyl-b-naphthylthiazole,naphth 1,2-d thiazole, 2-methyl,2-methyl-.beta.-naphthothiazole,2-methyinaphtho 1,2-d thiazole,2-methylbenzo e 1,3 benzothiazole
75892
2-methylbenzo[e][1,3]benzothiazole
CC1=NC2=C(S1)C=CC3=CC=CC=C32
2682-45-3
C12H9NS
199.271
MFCD00004951
OUXMJRMYZCEVKO-UHFFFAOYSA-N
2-methylnaphtho 1,2-d thiazole,2-methyl-beta-naphthothiazole,2-methylnaphtho 1,2-d 1,3 thiazole,naphtho 1,2-d thiazole, 2-methyl,2-methyl-b-naphthylthiazole,naphth 1,2-d thiazole, 2-methyl,2-methyl-.beta.-naphthothiazole,2-methyinaphtho 1,2-d thiazole,2-methylbenzo e 1,3 benzothiazole
75892
2-methylbenzo[e][1,3]benzothiazole
CC1=NC2=C(S1)C=CC3=CC=CC=C32
Boiling Point 168°C
Quantity 10g
Melting Point 96°C
Color Yellow
Formula Weight 199.27
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Methylnaphtho[1,2-d]thiazole

EINECSNumber : (5)-3439

TSCA : Yes