Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylnaphtho[2,3-d]oxazole, 98%, Alfa Aesar™

Click to view available options
Quantity:
5g
25g
20686-66-2
C12H9NO
183.21
MFCD00624597
WMIHEWJRNFUVNR-UHFFFAOYSA-N
2-methylnaphth 2,3-d oxazole,2-methylnaphtho 2,3-d oxazole,2-methylbenzo f 1,3 benzoxazole,2-methyl-naphtho 2,3-d oxazole,2-methylnaphtho 2,3-d 1,3 oxazole,2-methylbenzo f benzoxazole,pubchem8702
140743
2-methylbenzo[f][1,3]benzoxazole
CC1=NC2=CC3=CC=CC=C3C=C2O1
20686-66-2
C12H9NO
183.21
MFCD00624597
WMIHEWJRNFUVNR-UHFFFAOYSA-N
2-methylnaphth 2,3-d oxazole,2-methylnaphtho 2,3-d oxazole,2-methylbenzo f 1,3 benzoxazole,2-methyl-naphtho 2,3-d oxazole,2-methylnaphtho 2,3-d 1,3 oxazole,2-methylbenzo f benzoxazole,pubchem8702
140743
2-methylbenzo[f][1,3]benzoxazole
CC1=NC2=CC3=CC=CC=C3C=C2O1
Boiling Point 309°C to 311°C
Melting Point 85°C to 88°C
Quantity 5g
Formula Weight 183.21
Percent Purity 98%
Chemical Name or Material 2-Methylnaphtho[2,3-d]oxazole