Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylphenethyl alcohol, 98%, Acros Organics

Click to view available options
Quantity:
1g
19819-98-8
C9H12O
136.194
MFCD00044241
RUGISKODRCWQNE-UHFFFAOYSA-N
2-o-tolyl ethanol,2-methylphenethyl alcohol,2-2-methylphenyl ethanol,benzeneethanol, 2-methyl,o-methylphenethyl alcohol,phenethyl alcohol, o-methyl,2-methylbenzeneethanol,toluene-ethanol,unii-vx8700jgjp,2-2-methylphenyl ethan-1-ol
88267
2-(2-methylphenyl)ethanol
CC1=CC=CC=C1CCO
19819-98-8
C9H12O
136.194
MFCD00044241
RUGISKODRCWQNE-UHFFFAOYSA-N
2-o-tolyl ethanol,2-methylphenethyl alcohol,2-2-methylphenyl ethanol,benzeneethanol, 2-methyl,o-methylphenethyl alcohol,phenethyl alcohol, o-methyl,2-methylbenzeneethanol,toluene-ethanol,unii-vx8700jgjp,2-2-methylphenyl ethan-1-ol
88267
2-(2-methylphenyl)ethanol
CC1=CC=CC=C1CCO
Color Colorless
Refractive Index 1.5340 to 1.5360 (20°C, 589nm)
Quantity 1g
Infrared Spectrum Authentic
Density 1.01g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C6H4CH2CH2OH
Beilstein 06, 508
Formula Weight 136.19
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-Methylphenethyl alcohol, 98%
Show More Show Less