Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylphenylacetone, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
51052-00-7
C10H12O
148.205
MFCD00210465
WRLPDLDAOMBERO-UHFFFAOYSA-N
1-o-tolyl propan-2-one,1-2-methylphenyl propan-2-one,2-methylphenylacetone,2-methyphenyl acetone,alpha-methylphenylacetone,2-methylphenyl acetone,2-2-oxopropyl toluene,1-o-tolyl-propan-2-one,1-2-methylphenyl acetone
2734109
1-(2-methylphenyl)propan-2-one
CC1=CC=CC=C1CC(=O)C
51052-00-7
C10H12O
148.205
MFCD00210465
WRLPDLDAOMBERO-UHFFFAOYSA-N
1-o-tolyl propan-2-one,1-2-methylphenyl propan-2-one,2-methylphenylacetone,2-methyphenyl acetone,alpha-methylphenylacetone,2-methylphenyl acetone,2-2-oxopropyl toluene,1-o-tolyl-propan-2-one,1-2-methylphenyl acetone
2734109
1-(2-methylphenyl)propan-2-one
CC1=CC=CC=C1CC(=O)C
Refractive Index 1.5185 to 1.5205 (20°C, 589nm)
Packaging Glass bottle
Color Yellow
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 07, IV, 721
Formula Weight 148.21
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 2-Methylphenylacetone, 97%
Show More Show Less