Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (+/-)-2-Methylpiperazine, 98%

Click to view available options
Quantity:
25g
100g
109-07-9
C5H12N2
100.165
MFCD00005954
JOMNTHCQHJPVAZ-UHFFFAOYSA-N
piperazine, 2-methyl,2-methyl-piperazine,+/--2-methylpiperazine,2methylpiperazine,2-methylpiperazin,2-methypiperazine,2-methylpiperizine,3-methylpiperazine,2-methyl piperazine,pubchem8560
66057
2-methylpiperazine
CC1CNCCN1
109-07-9
C5H12N2
100.165
MFCD00005954
JOMNTHCQHJPVAZ-UHFFFAOYSA-N
piperazine, 2-methyl,2-methyl-piperazine,+/--2-methylpiperazine,2methylpiperazine,2-methylpiperazin,2-methypiperazine,2-methylpiperizine,3-methylpiperazine,2-methyl piperazine,pubchem8560
66057
2-methylpiperazine
CC1CNCCN1
Sensitivity Air sensitive; Hygroscopic
Melting Point 62°C to 66°C
Odor Amine-like
Quantity 100g
Flash Point 65°C (149°F)
Boiling Point 155°C to 156°C
UN Number UN2925
Beilstein 383566
Formula Weight 100.17
Percent Purity 98%
Chemical Name or Material (+/-)-2-Methylpiperazine
Show More Show Less

GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.

P210-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H228-H314

DOTInformation : Hazard Class: 4.1; Packaging Group: II

EINECSNumber : 203-644-2

RTECSNumber : TM1225020

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon