Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylpiperazine, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
109-07-9
C5H12N2
100.16
JOMNTHCQHJPVAZ-UHFFFAOYSA-N
piperazine, 2-methyl,2-methyl-piperazine,+/--2-methylpiperazine,2methylpiperazine,2-methylpiperazin,2-methypiperazine,2-methylpiperizine,3-methylpiperazine,2-methyl piperazine,pubchem8560
66057
2-methylpiperazine
CC1CNCCN1
109-07-9
C5H12N2
100.16
JOMNTHCQHJPVAZ-UHFFFAOYSA-N
piperazine, 2-methyl,2-methyl-piperazine,+/--2-methylpiperazine,2methylpiperazine,2-methylpiperazin,2-methypiperazine,2-methylpiperizine,3-methylpiperazine,2-methyl piperazine,pubchem8560
66057
2-methylpiperazine
CC1CNCCN1
Infrared Spectrum Authentic
Boiling Point 154.0°C to 156.0°C
Melting Point 61.0°C to 66.0°C
Quantity 2.5kg
Flash Point 63°C
Packaging Glass bottle
Color White to Yellow
Assay Percent Range 98%
Beilstein 23,17
Solubility Information Solubility in water: 78% (25°C).
Formula Weight 100.16
Physical Form Crystalline Powder, Crystals and/or Chunks
Percent Purity 98%
Chemical Name or Material 2-Methylpiperazine
Show More Show Less