Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methylpyridine-5-boronic acid, 97%

Click to view available options
Quantity:
250mg
1g
5g
659742-21-9
C6H8BNO2
136.945
MFCD06801736
MZUSCPDSQJSBSY-UHFFFAOYSA-N
6-methylpyridine-3-boronic acid,2-methylpyridine-5-boronic acid,6-methylpyridin-3-yl boronic acid,2-methyl-5-pyridinylboronic acid,2-picoline-5-boronic acid,2-methyl-5-boronopyridine,boronic acid, b-6-methyl-3-pyridinyl,6-methylpyridin-3-yl-3-boronic acid,6-methyl-3-pyridyl boronic acid,2-methylpyridin-5-ylboronic acid
2763081
(6-methylpyridin-3-yl)boronic acid
B(C1=CN=C(C=C1)C)(O)O
659742-21-9
C6H8BNO2
136.945
MFCD06801736
MZUSCPDSQJSBSY-UHFFFAOYSA-N
6-methylpyridine-3-boronic acid,2-methylpyridine-5-boronic acid,6-methylpyridin-3-yl boronic acid,2-methyl-5-pyridinylboronic acid,2-picoline-5-boronic acid,2-methyl-5-boronopyridine,boronic acid, b-6-methyl-3-pyridinyl,6-methylpyridin-3-yl-3-boronic acid,6-methyl-3-pyridyl boronic acid,2-methylpyridin-5-ylboronic acid
2763081
(6-methylpyridin-3-yl)boronic acid
B(C1=CN=C(C=C1)C)(O)O
Quantity 250mg
Color Brown to White
Formula Weight 136.94
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2-Methylpyridine-5-boronic acid