Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methylpyrimidine-5-carboxylic acid, 97%

Click to view available options
Quantity:
250mg
1g
5g
5194-32-1
C6H6N2O2
138.126
MFCD08235210
NMGIXZFBQPETOK-UHFFFAOYSA-N
2-methyl-5-pyrimidinecarboxylic acid,2-methylpyrimidine-5-carboxylicacid,5-pyrimidinecarboxylic acid, 2-methyl,2-methyl-5-pyrimidinecarboxylicacid,pubchem13290,acmc-209kwu,ksc269i7t,2-methyl-pyrimidine-5-carboxylic acid,5-pyrimidinecarboxylicacid, 2-methyl,2-methyl pyrimidine-5-carboxylic acid
13670875
2-methylpyrimidine-5-carboxylic acid
CC1=NC=C(C=N1)C(=O)O
5194-32-1
C6H6N2O2
138.126
MFCD08235210
NMGIXZFBQPETOK-UHFFFAOYSA-N
2-methyl-5-pyrimidinecarboxylic acid,2-methylpyrimidine-5-carboxylicacid,5-pyrimidinecarboxylic acid, 2-methyl,2-methyl-5-pyrimidinecarboxylicacid,pubchem13290,acmc-209kwu,ksc269i7t,2-methyl-pyrimidine-5-carboxylic acid,5-pyrimidinecarboxylicacid, 2-methyl,2-methyl pyrimidine-5-carboxylic acid
13670875
2-methylpyrimidine-5-carboxylic acid
CC1=NC=C(C=N1)C(=O)O
Applications
It is employed in the synthesis, spectral characterization and anticancer activity of new 2,3,6- substituted quinazolin-4(3h)-one derivatives.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Store in freezer at -20°C.
Quantity 5g
Melting Point 203°C to 205°C
Beilstein 508773
Formula Weight 138.13
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Methylpyrimidine-5-carboxylic acid

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

storageNote1 : -30° to -10°C