Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylpyrimidine, 98%, Thermo Scientific Chemicals

Catalog Number p-7077822
Click to view available options
Quantity:
1 g
5 g
5053-43-0
C5H6N2
94.11
LNJMHEJAYSYZKK-UHFFFAOYSA-N
pyrimidine, 2-methyl, 2-methyl pyrimidine, pubchem21974, 2-methyl-1,3-diazine, 2-methylpyrimidine, pyrimidine, 2-methyl-6ci,7ci,8ci,9ci
78748
2-methylpyrimidine
CC1=NC=CC=N1
5053-43-0
C5H6N2
94.11
LNJMHEJAYSYZKK-UHFFFAOYSA-N
pyrimidine, 2-methyl, 2-methyl pyrimidine, pubchem21974, 2-methyl-1,3-diazine, 2-methylpyrimidine, pyrimidine, 2-methyl-6ci,7ci,8ci,9ci
78748
2-methylpyrimidine
CC1=NC=CC=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -6.0°C
Density 1.0132g/mL
Boiling Point 130.0°C to 132.0°C
Flash Point 37.8°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.0132
Formula Weight 94.11
Percent Purity 98%
Chemical Name or Material 2-Methylpyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit