Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylquinoxaline 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
7251-61-8
C9H8N2
144.18
MFCD00006727
ALHUXMDEZNLFTA-UHFFFAOYSA-N
quinoxaline, 2-methyl,2-methyl quinoxaline,unii-03vu31mv6j,ccris 2948,methyl-quinoxaline,2-methyl-quinoxaline,acmc-1biyr,2-methylbenzopyrazine,2-methyl-1,4-naphthyridine,5-23-07-00219 beilstein handbook reference
23686
2-methylquinoxaline
CC1=NC2=CC=CC=C2N=C1
7251-61-8
C9H8N2
144.18
MFCD00006727
ALHUXMDEZNLFTA-UHFFFAOYSA-N
quinoxaline, 2-methyl,2-methyl quinoxaline,unii-03vu31mv6j,ccris 2948,methyl-quinoxaline,2-methyl-quinoxaline,acmc-1biyr,2-methylbenzopyrazine,2-methyl-1,4-naphthyridine,5-23-07-00219 beilstein handbook reference
23686
2-methylquinoxaline
CC1=NC2=CC=CC=C2N=C1
Boiling Point 245°C to 247°C
Flash Point 107°C
Refractive Index 1.6120 to 1.617
Specific Gravity 1.11
Infrared Spectrum Authentic
Density 1.1100g/mL
Quantity 5g
Packaging Glass bottle
Color Orange to Red
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 23, I, 44
Solubility Information Solubility in water: miscible. Other solubilities: miscible in ether,acetone,benzene,very soluble in ethanol,soluble in carbon tetrachloride
Formula Weight 144.18
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 2-Methylquinoxaline, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 230-664-9

RTECSNumber : VD3450000

TSCA : TSCA

Recommended Storage : Product may darken on standing