Alfa Aesar™ 2-Methylresorcinol, 98%

Manufacturer: Alfa Aesar™ A1182130

Quantity 250g

 View more versions of this product

Catalog No. AAA1182130

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 608-25-3
    Molecular Formula C7H8O2
    Formula Weight 124.14
    MDL Number MFCD00002271
    UN Number UN2811
    Beilstein 2042177
    Synonym 2,6-Dihydroxytoluene
    Chemical Name or Material 2-Methylresorcinol

    Specifications

    Formula Weight 124.14
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym 2,6-Dihydroxytoluene
    Health Hazard 1 Danger
    Quantity 250g
    MDL Number MFCD00002271
    CAS 608-25-3
    Melting Point 118° to 122°C
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    Molecular Formula C7H8O2
    Beilstein 2042177
    Flash Point 135°C (275°F)
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    UN Number UN2811
    Boiling Point 264°C
    RTECS Number VH2009500
    Chemical Name or Material 2-Methylresorcinol
    Assay 98%

    Description

    Applications
    2-Methylresorcinol is used in the preparation of aromatic benziporphyrins, tripyrrane analogs and C-5-bromo-2-hydroxyphenylcalix[4]-2-methylresorcinarene. Further, it is employed in oxidative and non-oxidative hair dye formulations.

    Solubility
    Soluble in water and methanol.

    Notes
    Incompatible with acid chlorides, acid anhydrides and oxidizing agents

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit