Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylstyrene, 97%, stabilized, Acros Organics

Click to view available options
Quantity:
5mL
Packaging:
Glass bottle
611-15-4
C9H10
118.18
NVZWEEGUWXZOKI-UHFFFAOYSA-N
2-methylstyrene,2-vinyltoluene,o-vinyltoluene,o-methylstyrene,styrene, o-methyl,1-methyl-2-vinylbenzene,benzene, 1-ethenyl-2-methyl,ortho-methylstyrene,unii-556uo5cl44,1-ethenyl-2-methyl-benzene
11904
1-ethenyl-2-methylbenzene
CC1=CC=CC=C1C=C
611-15-4
C9H10
118.18
NVZWEEGUWXZOKI-UHFFFAOYSA-N
2-methylstyrene,2-vinyltoluene,o-vinyltoluene,o-methylstyrene,styrene, o-methyl,1-methyl-2-vinylbenzene,benzene, 1-ethenyl-2-methyl,ortho-methylstyrene,unii-556uo5cl44,1-ethenyl-2-methyl-benzene
11904
1-ethenyl-2-methylbenzene
CC1=CC=CC=C1C=C
Melting Point -69°C
Boiling Point 169°C to 171°C
Density 0.9140g/mL
Flash Point 58°C
Specific Gravity 0.914
Packaging Glass bottle
Quantity 5mL
Assay Percent Range 96% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 118.18
Percent Purity 97%
Chemical Name or Material 2-Methylstyrene
Show More Show Less

GHS H Statement
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the env

GHS Signal Word: Warning

EINECSNumber : 210-256-7