Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylsulfanylpyrimidine-4-carboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1126-44-9
C6H6N2O2S
170.19
MFCD09834808
IAGNLKODEFUQDV-UHFFFAOYSA-N
2-methylthio pyrimidine-4-carboxylic acid,2-methylsulfanyl-pyrimidine-4-carboxylic acid,2-methylthiopyrimidine-4-carboxylic acid,2-thiomethylpyrimidine-4-carboxylic acid,2-methylsulfanyl pyrimidine-4-carboxylic acid,2-methylsulfanyl-pyrimidine-4-carboxylicacid,2-methylthio-4-pyrimidinecarboxylic acid,2-thioformylpyrimidine-4-carboxylic acid,2-methylthio-4-pyrimidinecarboxylicacid,2-mthylsulfanyl-pyrimidine-4-carboxylic acid
295775
2-methylsulfanylpyrimidine-4-carboxylic acid
CSC1=NC=CC(=N1)C(=O)O
1126-44-9
C6H6N2O2S
170.19
MFCD09834808
IAGNLKODEFUQDV-UHFFFAOYSA-N
2-methylthio pyrimidine-4-carboxylic acid,2-methylsulfanyl-pyrimidine-4-carboxylic acid,2-methylthiopyrimidine-4-carboxylic acid,2-thiomethylpyrimidine-4-carboxylic acid,2-methylsulfanyl pyrimidine-4-carboxylic acid,2-methylsulfanyl-pyrimidine-4-carboxylicacid,2-methylthio-4-pyrimidinecarboxylic acid,2-thioformylpyrimidine-4-carboxylic acid,2-methylthio-4-pyrimidinecarboxylicacid,2-mthylsulfanyl-pyrimidine-4-carboxylic acid
295775
2-methylsulfanylpyrimidine-4-carboxylic acid
CSC1=NC=CC(=N1)C(=O)O
Infrared Spectrum Authentic
Melting Point 208°C to 209°C
Packaging Glass bottle
Quantity 5g
Color White to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 170.19
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Methylsulfanylpyrimidine-4-carboxylic acid
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning