Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylsulfonyl-4,6-dimethoxypyrimidine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass Bottle
113583-35-0
C7H10N2O4S
218.227
MFCD00672151
ITDVJJVNAASTRS-UHFFFAOYSA-N
4,6-dimethoxy-2-methylsulfonyl pyrimidine,2-methanesulfonyl-4,6-dimethoxypyrimidine,2-methylsulfonyl-4,6-dimethoxypyrimidine,2-methylsulfony-4,6-dimethoxypyridine,4,6-dimethoxy-2-methylsulfonyl-pyrimidine,2-methanesulfonyl-4,6-dimethoxy-pyrimidine,4,6-dimethoxy-2-methylsulphonyl pyrimidine,pyrimidine, 4,6-dimethoxy-2-methylsulfonyl,pubchem23351,maybridge1_008822
838363
4,6-dimethoxy-2-methylsulfonylpyrimidine
COC1=CC(=NC(=N1)S(=O)(=O)C)OC
113583-35-0
C7H10N2O4S
218.227
MFCD00672151
ITDVJJVNAASTRS-UHFFFAOYSA-N
4,6-dimethoxy-2-methylsulfonyl pyrimidine,2-methanesulfonyl-4,6-dimethoxypyrimidine,2-methylsulfonyl-4,6-dimethoxypyrimidine,2-methylsulfony-4,6-dimethoxypyridine,4,6-dimethoxy-2-methylsulfonyl-pyrimidine,2-methanesulfonyl-4,6-dimethoxy-pyrimidine,4,6-dimethoxy-2-methylsulphonyl pyrimidine,pyrimidine, 4,6-dimethoxy-2-methylsulfonyl,pubchem23351,maybridge1_008822
838363
4,6-dimethoxy-2-methylsulfonylpyrimidine
COC1=CC(=NC(=N1)S(=O)(=O)C)OC
Packaging Glass Bottle
Color White
Quantity 5g
Melting Point 127°C to 131°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 218.15
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-Methylsulfonyl-4, 6-dimethoxypyrimidine
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.