Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylsulfonyl)acetonitrile, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2274-42-2
C3H5NO2S
119.14
FOTRKCAZUSJCQD-UHFFFAOYSA-N
methylsulfonylacetonitrile,2-methylsulfonyl acetonitrile,methylsulfonyl acetonitrile,methanesulfonylacetonitrile,mesylacetonitrile,2-methanesulfonylacetonitrile,cyanomethylsulfonylmethane,methanesulphonylacetonitrile,acetonitrile, methylsulfonyl,cyanomethyl methyl sulphone
75283
CHEBI:32444
2-methylsulfonylacetonitrile
CS(=O)(=O)CC#N
2274-42-2
C3H5NO2S
119.14
FOTRKCAZUSJCQD-UHFFFAOYSA-N
methylsulfonylacetonitrile,2-methylsulfonyl acetonitrile,methylsulfonyl acetonitrile,methanesulfonylacetonitrile,mesylacetonitrile,2-methanesulfonylacetonitrile,cyanomethylsulfonylmethane,methanesulphonylacetonitrile,acetonitrile, methylsulfonyl,cyanomethyl methyl sulphone
75283
CHEBI:32444
2-methylsulfonylacetonitrile
CS(=O)(=O)CC#N
Melting Point 83.0°C to 86.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Elemental Analysis 29.03 to 31.45% C, 11.29 to 12.23% N
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 119.14
Physical Form Powder or Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-(Methylsulfonyl)acetonitrile
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Danger

EINECSNumber : 218-888-5