Alfa Aesar™ 2-(Methylsulfonyl)benzenesulfonyl chloride, 95%

Manufacturer: Alfa Aesar™ H2736503

Quantity 1g

 View more versions of this product

Catalog No. AAH2736503

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 89265-35-0
    Molecular Formula C7H7ClO4S2
    Formula Weight 254.71
    MDL Number MFCD00216493
    UN Number UN3261
    Synonym 2-(Methanesulfonyl)benzenesulfonyl chloride
    Chemical Name or Material 2-(Methylsulfonyl)benzenesulfonyl chloride

    Specifications

    Formula Weight 254.71
    Molecular Formula C7H7ClO4S2
    UN Number UN3261
    Synonym 2-(Methanesulfonyl)benzenesulfonyl chloride
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 136° to 138°C
    MDL Number MFCD00216493
    CAS 89265-35-0
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Quantity 1g
    Chemical Name or Material 2-(Methylsulfonyl)benzenesulfonyl chloride
    Assay 95%

    Description

    Applications
    It finds its application in the synthesis and biological evaluation of inhibitors of thymidine monophosphate kinase from bacillus anthracis.

    Solubility
    Reacts with water.

    Notes
    Moisture sensitive. Store away from water/moisture, strong bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit