Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyltetrahydrofuran-3-thiol, 97%, mixed isomers, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
57124-87-5
C5H10OS
118.2
MFCD01632596
DBPHPBLAKVZXOY-UHFFFAOYSA-N
2-methyl-3-tetrahydrofuranthiol,2-methyltetrahydrofuran-3-thiol,tetrahydro-2-methyl-3-furanthiol,3-furanthiol, tetrahydro-2-methyl,2-methyl-3-mercaptotetrahydrofuran,pentitol, 1,4-anhydro-2,5-dideoxy-3-thio,3-mercapto-2-methyltetrahydrofuran,pubchem9972,ksc201q4p,2-methyl-3-tetrahydrofuran thiol
62128
2-methyloxolane-3-thiol
CC1C(CCO1)S
57124-87-5
C5H10OS
118.2
MFCD01632596
DBPHPBLAKVZXOY-UHFFFAOYSA-N
2-methyl-3-tetrahydrofuranthiol,2-methyltetrahydrofuran-3-thiol,tetrahydro-2-methyl-3-furanthiol,3-furanthiol, tetrahydro-2-methyl,2-methyl-3-mercaptotetrahydrofuran,pentitol, 1,4-anhydro-2,5-dideoxy-3-thio,3-mercapto-2-methyltetrahydrofuran,pubchem9972,ksc201q4p,2-methyl-3-tetrahydrofuran thiol
62128
2-methyloxolane-3-thiol
CC1C(CCO1)S
Infrared Spectrum Authentic
Refractive Index 1.4730 to 1.4910
Packaging Glass bottle
Color Colorless to Yellow
Density 1.0400g/mL
Quantity 5g
Flash Point 40°C
Specific Gravity 1.04
Boiling Point 160.0°C to 180.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 118.2
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Methyltetrahydrofuran-3-thiol, Mixture of isomers
Show More Show Less

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning