Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyltetrahydrofuran, ReagentPlus™, ≥99.5%, Honeywell Burdick & Jackson™

Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
4 x 4 L
Packaging:
Glass Bottle
96-47-9
C5H10O
86.134
MFCD00005367
JWUJQDFVADABEY-UHFFFAOYSA-N
2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
7301
2-methyloxolane
CC1CCCO1
96-47-9
C5H10O
86.134
MFCD00005367
JWUJQDFVADABEY-UHFFFAOYSA-N
2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
7301
2-methyloxolane
CC1CCCO1

Reagent Grade, contains 150-400 ppm BHT as stabilizer,≥99.5%

  • ≥95% for general laboratory use.
Melting Point <-20°C
Color Colorless
UN Number UN2536
Beilstein 102448
Formula Weight 86.13g/mol
Percent Purity ≥99.5%
Grade ReagentPlus™
Density 0.86g/mL at 25°C
Boiling Point 78°C to 80°C
Quantity 500 mL
Flash Point −10°C (14°F)
Packaging Glass Bottle
Refractive Index n20/D 1.406(lit.)
Chemical Name or Material 2-Methyltetrahydrofuran
Show More Show Less

P210-P280-P301 + P312 + P330-P305 + P351 + P338 + P310-P370 + P378-P403 + P235

H225-H302-H315-H318

DOTInformation : 3/PGII

EINECSNumber : 202-507-4

RTECSNumber : LU2800000