Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyltetrahydrofuran, Spectrum™

Click to view available options
Quantity:
100 mL
1 L
4 L
200 L
Packaging:
Amber Glass Bottle
Drum
96-47-9
C5H10O
86.13
JWUJQDFVADABEY-UHFFFAOYNA-N
2-methyloxolane
CC1CCCO1
96-47-9
C5H10O
86.13
JWUJQDFVADABEY-UHFFFAOYNA-N
2-methyloxolane
CC1CCCO1

M2406, 96-47-9, C5H10O

Spectrum™ 2-Methyltetrahydrofuran
Packaging Amber Glass Bottle
Quantity 4 L
CAS Max % 100%
UN Number UN2536