Alfa Aesar™ 2-(Methylthio)benzimidazole, 98+%

Manufacturer: Alfa Aesar™ A1068618

Quantity 50g

 View more versions of this product

Catalog No. AAA1068618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7152-24-1
    Molecular Formula C8H8N2S
    Formula Weight 164.23
    MDL Number MFCD00005594
    Beilstein 122360
    Synonym 2-(Methylmercapto)benzimidazole
    Chemical Name or Material 2- (Methylthio)benzimidazole

    Specifications

    Formula Weight 164.23
    Molecular Formula C8H8N2S
    MDL Number MFCD00005594
    CAS 7152-24-1
    Synonym 2-(Methylmercapto)benzimidazole
    TSCA TSCA
    Beilstein 122360
    Melting Point 203° to 205°C
    Quantity 50g
    RTECS Number DD9480000
    Chemical Name or Material 2- (Methylthio)benzimidazole
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit