Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Methylthio)naphthalene, 98%

Manufacturer:  Alfa Aesar™ B2236814

Quantity 25g

 View more versions of this product

Catalog No. AAB2236814


  Check Availability
Add to cart

Specifications

Description

Applications
2-(Methylthio)naphthalene is used to produce 1-chloro-2-methylsulfanylnaphthalene. This reaction will need reagent CuCl2-Al2O3, and the menstruum benzene

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

7433-79-6
174.27
1859409
2-(Methylthio)naphthalene
C11H10S
MFCD00051615
2-(Methylmercapto)naphthalene

Specifications

7433-79-6
174.27
1859409
62° to 63°C
Stench
98%
C11H10S
MFCD00051615
2-(Methylmercapto)naphthalene
25g
2-(Methylthio)naphthalene
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit